Learn More
MMP3 Recombinant Rabbit Monoclonal Antibody (45H6L22), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marque: Invitrogen 701285
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.01000kg
Description
This antibody is predicted to react with equine, non-human primate, rabbit and porcine based on sequence homology. Intact IgG appears on a non-reducing gel as ∽150 kDa band and upon reduction generating a ∽25 kDa light chain band and a ∽50 kDa heavy chain. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
MMP3 Recombinant Monoclonal antibody specifically detects MMP3 in Human samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spécification
MMP3 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
CHDS6; EC 3.4.24.17; EMS-2; matrix metallo protease; matrix metallopeptidase 3; matrix metallopeptidase 3 (stromelysin 1, progelatinase); matrix metalloproteinase 3; matrix metalloproteinase 3 (stromelysin 1, progelatinase); matrix metalloproteinase 3 precursor; matrix metalloproteinase-3; metalloproteinase 3 receptor; MMP; MMP10; MMP3; MMP-3; MMP-3 protein; MMPs; progelatinase; proteoglycanase; SL-1; SLN1; SLN-1; STMY; STMY1; Str1; STR-1; stromelysin; stromelysin 1; Stromelysin1; stromelysin-1; Transin1; Transin-1 | |
Recombinant protein corresponding to amino acids 255–477 of human MMP-3 | |
100 μg | |
Primary | |
Human | |
Liquid |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
45H6L22 | |
Unconjugated | |
P08254 | |
Rabbit | |
Protein A | |
RUO | |
4314 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |