missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEL, Mouse, Peroxidase, Clone: KNH-30, Abnova™
Mouse monoclonal antibody raised against recombinant CEL.
Marque: Abnova MAB6595.50ug
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.00010kg
Spécification
CEL | |
Monoclonal | |
Peroxidase | |
ELISA (0.1-1 ug/mL) Immunohistochemistry (5-10 ug/mL) The optimal working dilution should be determined by the end user. | |
Mouse | |
50 μg | |
Primary | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Immunohistochemistry | |
KNH-30 | |
Mouse monoclonal antibody raised against recombinant CEL. | |
In serum (2% Block Ace, 0.1% proclin) | |
Recombinant protein (conjugated with BSA) corresponding to CEL. | |
RUO | |
Human, Mouse, Rat | |
Liquid |