Learn More
CLOCK, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant CLOCK.
Marque: Abnova H00009575-A01.50uL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes a protein that belongs to the basic helix-loop-helix (bHLH) family of transcription factors. Polymorphisms within the encoded protein have been associated with circadian rhythm sleep disorders. A similar protein in mice is a circadian regulator that acts as a transcription factor and forms a heterodimer with aryl hydrocarbon receptor nuclear translocator-like to activate transcription of mouse period 1. [provided by RefSeq
Sequence: PVMSQATNLPIPQGMSQFQFSAQLGAMQHLKDQLEQRTRMIEANIHRQQEELRKIQEQLQMVHGQGLQMFLQQSNPGLNFGSVQLSSGNSSNIQQLAPINSpécification
CLOCK | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant CLOCK. | |
CLOCK | |
KAT13D/KIAA0334/bHLHe8 | |
Mouse | |
50 μL | |
Primary | |
Human | |
Antisera |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_004898 | |
CLOCK | |
CLOCK (NP_004889, 497 a.a. to 596 a.a) partial recombinant protein with GST tag. | |
RUO | |
9575 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
For Research Use Only