missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Integrin alpha 2/CD49b Antibody, R&D Systems™

Rat Monoclonal Antibody
Marque: R&D Systems MAB1740-SP
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Integrin alpha 2/CD49b Monoclonal specifically detects Integrin alpha 2/CD49b in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.
Spécification
| Integrin alpha 2/CD49b | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 2.5 ug/10^6 cells | |
| Q62469 | |
| ITGA2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Integrin alpha 2/CD49b Tyr27-Thr1129 Accession # Q62469 | |
| 25 μg | |
| Primary | |
| Detects mouse Integrin alpha 2/CD49b in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse Integrin alpha 3, alpha 4, alpha 5, alpha E, or alpha V is observed. | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| 235033 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| alpha-2 subunit, CD49b, CD49b antigen, integrin, alpha 2 (CD49B, alpha 2 subunit of VLA-2 receptor), ITGA2, VLA 2, VLA-2 alpha | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3673 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu