missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Integrin alpha 2/CD49b Antibody, R&D Systems™

Rat Monoclonal Antibody
201.00€ - 406.00€
Spécification
| Antigène | Integrin alpha 2/CD49b |
|---|---|
| Clone | 235033 |
| Concentration | LYOPH |
| Dilution | Flow Cytometry 2.5 ug/10^6 cells |
| Applications | Flow Cytometry |
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
18739688
![]() |
R&D Systems
MAB1740-SP |
25 μg |
201.00€
25µg |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
|
18180951
|
R&D Systems
MAB1740 |
100 μg |
406.00€
100µg |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
Description
Integrin alpha 2/CD49b Monoclonal specifically detects Integrin alpha 2/CD49b in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.Spécification
| Integrin alpha 2/CD49b | |
| LYOPH | |
| Flow Cytometry | |
| Unconjugated | |
| Rat | |
| Mouse | |
| Q62469 | |
| 3673 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Integrin alpha 2/CD49b Tyr27-Thr1129 Accession # Q62469 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse Integrin alpha 2/CD49b in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse Integrin alpha 3, alpha 4, alpha 5, alpha E, or alpha V is observed. |
| 235033 | |
| Flow Cytometry 2.5 ug/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| alpha-2 subunit, CD49b, CD49b antigen, integrin, alpha 2 (CD49B, alpha 2 subunit of VLA-2 receptor), ITGA2, VLA 2, VLA-2 alpha | |
| ITGA2 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit