missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Mouse LIX (CXCL6) (70 aa) Recombinant Protein, PeproTech®
Code nomenclature Nacres: NA.28
Click to view available options
Quantité:
1 mg
100 μg
20 μg
250 μg
5 μg
500 μg
Conditionnement:
100µg
1mg
20µg
250µg
500µg
5µg
Description
Recombinant Murine LIX is a 7.6 kDa protein consisting of amino acid residues 9-78 of the full length LIX protein (70 amino acids). This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
CXCL6 is a chemotactic for neutrophil granulocytes. It signals through binding and activation of its receptors (CXCR1 and CXCR2). In addition to its chemotactic and angiogenic properties, it has strong antibacterial activity against Gram-positive and Gram-negative bacteria (90-fold-higher when compared to CXCL5 and CXCL7).
Spécification
Spécification
| À utiliser avec (application) | Functional Assay |
| Formule | protein with no preservative |
| Poids moléculaire | 7.6 kDa |
| Nom | Mouse LIX (CXCL6) (70 aa) |
| Quantité | 100 μg |
| Source | E. coli |
| État réglementaire | RUO |
| Concentration en endotoxines | <1 EU/ μg |
| Alias de gène | C Cmotif chemokine; C x C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 5; chemokine (C-X-C motif) ligand 6; chemokine (C-X-C motif) ligand 6 (granulocyte chemotactic protein 2); chemokine alpha 3; CKA3; CKA-3; CXC; CXC chemokine LIX; CXC motif chemokine; C-X-C motif chemokine 5; C-X-C motif chemokine 6; C-X-C motif chemokine ligand 6; CXCL; Cxcl5; CXCL6; Cytokine LIX; GCP2; GCP-2; granulocyte chemotactic protein 2; Scyb5; SCYB6; small inducible cytokine subfamily B (Cys-X-Cys), member 6 (granulocyte chemotactic protein 2); Small inducible cytokine subfamily B (Cys-X-Cys), member b; small-inducible cytokine B5; Small-inducible cytokine B6; Small-inducible cytokine B6, N-processed variant 1; Small-inducible cytokine B6, N-processed variant 2; Small-inducible cytokine B6, N-processed variant 3 |
| Nom usuel | CXCL6 |
| Afficher plus |
Suggestions de produits
Clients qui ont consulté cet article ont également consulté
Viewing 1-4 of
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Gibco™ Mouse LIX (CXCL6) (70 aa) Recombinant Protein, PeproTech® >
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
