missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse/Rat CD31/PECAM-1 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 16 publications
Marque: R&D Systems FAB3628G-100
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
CD31/PECAM-1 Polyclonal specifically detects CD31/PECAM-1 in Mouse, Rat samples. It is validated for Flow Cytometry.
Spécification
| CD31/PECAM-1 | |
| Polyclonal | |
| Flow Cytometry 5 uL/10^6 cells | |
| Q08481 | |
| PECAM1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse CD31/PECAM-1 Glu18-Lys590 Accession # Q08481 | |
| 100 Tests | |
| Primary | |
| Detects mouse CD31/PECAM-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant human CD31 and recombinant porcine CD31 is observed. Detects mouse CD31 and rat CD31 in flow cytometry. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
| Flow Cytometry | |
| Alexa Fluor 488 | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| adhesion molecule, CD31, CD31 antigen, CD31/EndoCAM, EndoCAM, FLJ34100, FLJ58394, GPIIA', PECA1, PECAM1, PECAM-1, PECAM-1, CD31/EndoCAM, platelet endothelial cell adhesion molecule, platelet endothelial cell adhesion molecule-1, platelet/endothelial cell adhesion molecule | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 5175 | |
| Mouse, Rat | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu