Learn More
Mouse u-Plasminogen Activator (uPA)/Urokinase Antibody, R&D Systems™
Rat Monoclonal Antibody
Marque: R&D Systems MAB9185-SP
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
u-Plasminogen Activator/Urokinase Monoclonal specifically detects u-Plasminogen Activator/Urokinase in Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Spécification
| u-Plasminogen Activator/Urokinase | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| ATF, EC 3.4.21, EC 3.4.21.73, plasminogen activator, urokinase, PLAU, uPA, u-PA, uPlasminogen Activator, U-plasminogen activator, urinary, Urokinase, urokinase-type plasminogen activator | |
| Mouse myeloma cell line NS0-derived recombinant mouse uPA Met1-Phe433 Accession # P06869 | |
| 25 μg | |
| Primary | |
| Detects mouse uPA in direct ELISAs and Western blots. | |
| Mouse | |
| IgG2a |
| Western Blot, Immunohistochemistry | |
| 901420 | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-25 ug/mL | |
| P06869 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5328 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |