Learn More
Mouse u-Plasminogen Activator (uPA)/Urokinase Antibody, R&D Systems™
Rat Monoclonal Antibody
210.00€ - 512.00€
Spécification
| Antigène | u-Plasminogen Activator/Urokinase |
|---|---|
| Clone | 901420 |
| Dilution | Western Blot 1 ug/mL, Immunohistochemistry 5-25 ug/mL |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
18799208
![]() |
R&D Systems
MAB9185-SP |
25 μg |
210.00€
25µg |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
18793869
![]() |
R&D Systems
MAB9185-100 |
100 μg |
512.00€
100µg |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
Description
u-Plasminogen Activator/Urokinase Monoclonal specifically detects u-Plasminogen Activator/Urokinase in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Spécification
| u-Plasminogen Activator/Urokinase | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-25 ug/mL | |
| Monoclonal | |
| Rat | |
| Mouse | |
| P06869 | |
| 5328 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 901420 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| ATF, EC 3.4.21, EC 3.4.21.73, plasminogen activator, urokinase, PLAU, uPA, u-PA, uPlasminogen Activator, U-plasminogen activator, urinary, Urokinase, urokinase-type plasminogen activator | |
| Mouse myeloma cell line NS0-derived recombinant mouse uPA Met1-Phe433 Accession # P06869 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse uPA in direct ELISAs and Western blots. |