missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPL37 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 15190-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit.Spécification
MRPL37 | |
Polyclonal | |
Unconjugated | |
MRPL37 | |
L2mt, L37mt, MRP L2, MRP L37, MRPL2, MRPL37, RPML2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
51253, 56280, 56281 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6AXT0, Q921S7, Q9BZE1 | |
MRPL37 | |
MRPL37 Fusion Protein Ag7276 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |