missing translation for 'onlineSavingsMsg'
Learn More

MRPL47 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 24728-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16833215

  • 399.00€ / 150µL
Expédition estimée: 29-04-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. This gene is immediately adjacent to the gene for BAF complex 53 kDa subunit protein a (BAF53a), in a tail-to-tail orientation. Two transcript variants encoding different protein isoforms have been identified.
Spécification

Spécification

MRPL47
Polyclonal
Unconjugated
MRPL47
CGI 204, L47mt, MRP L47, MRPL47, NCM1
Rabbit
Antigen Affinity Chromatography
RUO
57129, 74600
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.48 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8K2Y7, Q9HD33
MRPL47
MRPL47 Fusion Protein Ag11744
150 μL
Primary
Human, Mouse
Antibody
IgG
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions