Learn More
MRPL48 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 14677-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Code produit. 16833434
  • 325.38€ / 150µL
Expédition estimée: 06-10-2022
Description
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. A pseudogene corresponding to this gene is found on chromosome 6p.
Spécification
Spécification
MRPL48
Polyclonal
Unconjugated
MRPL48
CGI 118, HSPC290, L48MT, MRP L48, MRPL48
Rabbit
Antigen Affinity Chromatography
RUO
293149, 51642, 52443
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry
0.42 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8JZS9, Q96GC5
MRPL48
MRPL48 Fusion Protein Ag6387
150 μL
Primary
Rat, Mouse, Human