Learn More
MS4A2 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 26808-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The MS4A2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human MS4A2. This antibody recognizes human, mouse antigen. The MS4A2 antibody has been validated for the following applications: WB, ELISA analysis.
Spécification
| MS4A2 | |
| 550 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| APY, ATOPY, FCER1B, FCERI, IgE Fc receptor subunit beta, IGEL, IGER, IGHER, MS4A1, MS4A2 | |
| 20 μL | |
| 2206 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |