Learn More
MUS81 Mouse, anti-Human, Unconjugated, Clone: 2D9B7, Proteintech
Mouse Monoclonal Antibody
Marque:  Proteintech 67351-1-IG-150UL
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.00020kg
 Afficher plus de versions de ce produit
Code produit. 17281466
  • 399.00€ / 150µL
Expédition estimée: 22-08-2024
pour voir le stock disponible 
Description
Description
The MUS81 antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human MUS81. This antibody recognizes human antigen. The MUS81 antibody has been validated for the following applications: WB,ELISA analysis.
Spécification
Spécification
MUS81
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, ELISA
2D9B7
Unconjugated
150 μL
80198