missing translation for 'onlineSavingsMsg'
Learn More

Muskelin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 14735-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16893524

  • 399.00€ / 150µL
Expédition estimée: 28-05-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

MAP kinase-interacting serine/threonine kinases (Mnk) function in signal transduction pathways through phosphorylation of eIF4E and are directly phosphorylated by ERK or p38 MAP kinases. In human and mouse there are two Mnk genes. Mnk1 is activated by phosphorylation at Thr197 and Thr202. Mnk1 and Mnk2 are essential for the phosphorylation of eIF4E at Ser209 however they differ markedly in their activity and regulatory patterns. Mnk1 is responsible for inducible phosphorylation of eIF4E while Mnk2 is responsible for basal levels of phosphorylation. Mnk1 has been implicated in signaling in response to oxidative stress by increasing eIF4E phosphorylation upon H2O2 treatment in mice. Oxidative stress induced Mnk1 activity has also been linked to hyperproliferative diseases through the over-activation of eIF4E. Reactivity with Mnk2 has not been tested.
Spécification

Spécification

Muskelin
Polyclonal
Unconjugated
MKLN1
MKLN1, Muskelin, TWA2
Rabbit
Antigen Affinity Chromatography
RUO
27418, 4289, 83536
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.15 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O89050, Q99PV3, Q9UL63
MKLN1
Muskelin Fusion Protein Ag6147
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions