missing translation for 'onlineSavingsMsg'
Learn More

MYBBP1A Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 14524-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16833204

  • 121.38€ / 20µL
Expédition estimée: 28-09-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The Myb family of transcription factors, which includes the structurally related A-, B-, and c-Myb genes, regulate differentiation, cellular growth and transactivating gene expression. c-Myb plays an essential role in controlling the proliferation and differentiation of hematopoietic cells, cellular levels of c-Myb decreasing as cells reach terminal differentiation. Myb-binding protein 1A (MYBBP1A) is a novel nuclear protein localized predomitly, but not exclusively, in nucleoli. Although initially isolated as a c-Myb-interacting protein, MYBBP1A is expressed ubiquitously, and associates with a number of different transcription factors. MYBBP1A associates with the aromatic hydrocarbon receptor (AhR), enhancing transactivation and increasing AhR-dependent gene expression. MYBBP1A is a powerful negative regulator of PPAR gamma coactivator 1 alpha (PGC-1 alpha) by p38 MAPK and also acts as a nucleocytoplasmic shuttling protein that utilizes CRM1-dependent and independent nuclear export pathways.
Spécification

Spécification

MYBBP1A
Polyclonal
Unconjugated
MYBBP1A
MYB binding protein (P160) 1a, Myb binding protein 1A, MYBBP1A, P160, PAP2
Rabbit
Antigen Affinity Chromatography
RUO
10514
-20°C
Liquid
Immunoprecipitation, Flow Cytometry, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence
0.35 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9BQG0
MYBBP1A
MYBBP1A Fusion Protein Ag6008
20 μL
Primary
Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions