Learn More
Myelin Basic Protein Ab-1 Rabbit Polyclonal Antibody, Epredia™
Myelin Basic Protein Ab-1, Rabbit Polyclonal Antibody is one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available.
Marque: Epredia RB-1460-A1
Description
Myelin basic protein accounts for about 30% of the proteins in myelin from the central nervous system. It is best known for its capacity to induce experimental allergic encephalomyelitis (EAE), a T-lymphocyte mediated disease due to delayed-type hypersensitivity; though each animal species appears to respond to a different fragment of the 170 amino acid polypeptide. Myelin basic protein is expressed in oligodendrocytes, myelin of white matter in the brain and spinal cord and in peripheral nerves.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human. Others not known.
Immunogen: Myelin basic protein isolated from human brain
Positive Control: Brain
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin )
Spécification
| Myelin Basic Protein Ab-1 | |
| Polyclonal | |
| Rabbit | |
| 500 μL | |
| Neuroscience | |
| Human | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Myelin basic protein isolated from human brain | |
| IVD | |
| Primary | |
| Concentrated |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.