missing translation for 'onlineSavingsMsg'
Learn More

MYL12B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 10324-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16896633

  • 349.00€ / 150µL
Expédition estimée: 16-12-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This gene encodes a nonsarcomeric myosin regulatory light chain. This protein is activated by phosphorylation and regulates smooth muscle and non-muscle cell contraction. This protein may also be involved in DNA damage repair by sequestering the transcriptional regulator apoptosis-antagonizing transcription factor (AATF)/Che-1 which functions as a repressor of p53-driven apoptosis. Alternate splicing results in multiple transcript variants. A pseudogene of this gene is found on chromosome 8.
Spécification

Spécification

MYL12B
Polyclonal
Unconjugated
MYL12B
MLC 2, MLC 2A, MLC B, MLC20, MRLC2, MYL12B, MYLC2B, SHUJUN 1
Rabbit
Antigen Affinity Chromatography
RUO
103910, 50685, 67938
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O14950, P18666, Q3THE2
Myl12b
MYL12B Fusion Protein Ag0398
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions