Learn More
MYT1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 26204-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The protein encoded by this gene is a member of a family of neural specific, zinc finger-containing DNA-binding proteins. This kinase preferentially phosphorylates and inactivates cell division cycle 2 protein (CDC2), and thus negatively regulates cell cycle G2/M transition. This kinase is associated with the membrane throughout the cell cycle. Its activity is highly regulated during the cell cycle. Protein kinases AKT1/PKB and PLK (Polo-like kinase) have been shown to phosphorylate and regulate the activity of this kinase. Alternatively spliced transcript variants encoding distinct isoforms have been reported.The protein binds to the promoter regions of proteolipid proteins of the central nervous system and plays a role in the developing nervous system.Spécification
| MYT1 | |
| Polyclonal | |
| Unconjugated | |
| MYT1 | |
| C20orf36, KIAA0835, KIAA1050, MTF1, myelin transcription factor 1, Myelin transcription factor I, MYT1, MYTI, PLPB1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 4661 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q01538 | |
| MYT1 | |
| MYT1 Fusion Protein Ag24580 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |