missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYT1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Spécification
| Antigène | MYT1 |
|---|---|
| Concentration | 0.3 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugué | Unconjugated |
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
|
16844725
|
Proteintech
26204-1-AP-20UL |
20 μL |
145.00€
20µL |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
|
16834725
|
Proteintech
26204-1-AP-150UL |
150 μL |
410.00€
150µL |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
Description
The protein encoded by this gene is a member of a family of neural specific, zinc finger-containing DNA-binding proteins. This kinase preferentially phosphorylates and inactivates cell division cycle 2 protein (CDC2), and thus negatively regulates cell cycle G2/M transition. This kinase is associated with the membrane throughout the cell cycle. Its activity is highly regulated during the cell cycle. Protein kinases AKT1/PKB and PLK (Polo-like kinase) have been shown to phosphorylate and regulate the activity of this kinase. Alternatively spliced transcript variants encoding distinct isoforms have been reported.The protein binds to the promoter regions of proteolipid proteins of the central nervous system and plays a role in the developing nervous system.Spécification
| MYT1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q01538 | |
| 4661 | |
| MYT1 Fusion Protein Ag24580 | |
| Primary | |
| -20°C | |
| MYT1 |
| 0.3 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| C20orf36, KIAA0835, KIAA1050, MTF1, myelin transcription factor 1, Myelin transcription factor I, MYT1, MYTI, PLPB1 | |
| MYT1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit