missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NADK Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 15548-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
The human trans-activation response (TAR) RNA binding protein 1 (TRBP1) was initially identified as a protein that binds to the HIV-1 TAR RNA and activates the long terminal repeat (LTR) expression in the absence and presence of the viral trans-activator Tat. This binding is enhanced by the presence of co-factors such as elongation factor 1-a (EF-1-a), polypyrimidine tract-binding protein (PTB), and the chaperonin-like protein stimulator of TAR RNA-binding proteins (SRB). TRBP1 may act to disengage RNA polymerase II from TAR during transcriptional elongation if the RNA polymerase stalls at the TAR during transcriptional elongation.Spécification
NADK | |
Polyclonal | |
Unconjugated | |
NADK | |
NAD kinase, NADK, Poly(P)/ATP NAD kinase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100125370, 192185, 65220 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O95544, P58058 | |
NADK | |
NADK Fusion Protein Ag7906 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |