missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nav1.5 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque: Novus Biologicals NBP2-94760-0.1ml
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Nav1.5 Polyclonal antibody specifically detects Nav1.5 in Mouse samples. It is validated for Western Blot
Spécification
| Nav1.5 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000 | |
| cardiac sodium channel alpha subunit, cardiac tetrodotoxin-insensitive voltage-dependent sodium channel alpha subunit, CDCD2VF1, HB1sodium channel, voltage-gated, type V, alpha (long QT syndrome 3), HB2, HBBD, HH1CMD1E, ICCD, IVF, LQT3SSS1CMPD2, Nav1.5, PFHB1, Sodium channel protein cardiac muscle subunit alpha, sodium channel protein type 5 subunit alpha, sodium channel protein type V alpha subunit, Sodium channel protein type V subunit alpha, sodium channel, voltage-gated, type V, alpha subunit, Voltage-gated sodium channel subunit alpha Nav1.5 | |
| A synthetic peptide corresponding to a sequence within amino acids 1950-2016 of human Nav1.5 (NP_932173.1). YVMSENFSRPLGPPSSSSISSTSFPPSYDSVTRATSDNLQVRGSDYSHSEDLADFPPSPDRDRESIV | |
| 0.1 mL | |
| Cancer, Hypoxia | |
| 6331 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu