missing translation for 'onlineSavingsMsg'
Learn More

NCS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 10506-2-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16836933

  • 121.38€ / 20µL
Expédition estimée: 30-09-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This gene is a member of the neuronal calcium sensor gene family, which encode calcium-binding proteins expressed predomitly in neurons. The protein encoded by this gene regulates G protein-coupled receptor phosphorylation in a calcium-dependent manner and can substitute for calmodulin. The protein is associated with secretory granules and modulates synaptic transmission and synaptic plasticity. Multiple transcript variants encoding different isoforms have been found for this gene.
Spécification

Spécification

NCS1
Polyclonal
Unconjugated
NCS1
FLUP, FREQ, Frequenin homolog, frequenin homolog (Drosophila), Frequenin like protein, NCS 1, NCS1, Neuronal calcium sensor 1
Rabbit
Antigen Affinity Chromatography
RUO
14299, 23413, 65153
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.5 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P62166, P62168, Q8BNY6
Ncs1
NCS1 Fusion Protein Ag0784
20 μL
Primary
Human, Rat, Mouse
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions