missing translation for 'onlineSavingsMsg'
Learn More

NELFE Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 10705-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16827263

  • 325.38€ / 150µL
Expédition estimée: 03-10-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

10705-1-AP is obtained by immunizing animals with full-length RDBP of human origin.

RDBP is part of a complex termed negative elongation factor (NELF) which represses RNA polymerase II transcript elongation. This protein bears similarity to nuclear RNA-binding proteins; however, it has not been demonstrated that this protein binds RNA. The protein contains a tract of alternating basic and acidic residues, largely arginine (R) and aspartic acid (D). The protein encoded by this gene is part of a complex termed negative elongation factor (NELF) which represses RNA polymerase II transcript elongation. This protein bears similarity to nuclear RNA-binding proteins; however, it has not been demonstrated that this protein binds RNA. The protein contains a tract of alternating basic and acidic residues, largely arginine (R) and aspartic acid (D). The gene localizes to the major histocompatibility complex (MHC) class III region on chromosome 6.
Spécification

Spécification

NELFE
Polyclonal
Unconjugated
NELFE
D6S45, Negative elongation factor E, NELF E, NELFE, RD, RD RNA binding protein, RDBP, RDP, RNA binding protein RD
Rabbit
Antigen Affinity Chromatography
RUO
27632, 294258, 7936
-20°C
Liquid
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry
0.2 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P18615, P19426
Nelfe
NELFE Fusion Protein Ag1186
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions