missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nestin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 4 publications
Marque: Novus Biologicals NB300-265SS
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Nestin Polyclonal specifically detects Nestin in Human, Canine, Mouse (Negative), Rabbit (Negative), Rat (Negative) samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Spécification
| Nestin | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.1% Sodium Azide | |
| FLJ21841, nestin | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This is specific for human nestin. | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:100-1:500 | |
| P48681 | |
| NES | |
| Recombinant human Nestin protein (corresponding to residues 1464-1614). [UniProt# P48681] | |
| 0.025 mL | |
| Angiogenesis, Cancer, Cellular Markers, Cytoskeleton Markers, Extracellular Matrix, Neuronal Cell Markers, Neuronal Stem Cell Markers, Neuroscience, Stem Cell Markers | |
| 10763 | |
| Human, Canine, Mouse (Negative), Rabbit (Negative), Rat (Negative) | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu