missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFkB p105/p50, p Ser337 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque: Novus Biologicals NB100-82074
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
NFkB p105/p50 Polyclonal specifically detects NFkB p105/p50 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Spécification
| NFkB p105/p50 (p Ser337) | |
| Polyclonal | |
| Unconjugated | |
| PBS (without Mg2+ and Ca2+), pH 7.4, 150 mM NaCl, 50% glycerol with 0.02% Sodium Azide | |
| DKFZp686C01211, DNA binding factor KBF1, DNA-binding factor KBF1, EBP-1, KBF1, NF-kappaB, NF-kappa-B, NF-kappabeta, NFKB-p105, NFKB-p50, nuclear factor kappa-B DNA binding subunit, nuclear factor NF-kappa-B p105 subunit, nuclear factor NF-kappa-B p50 subunit, nuclear factor of kappa light polypeptide gene enhancer in B-cells 1MGC54151, p105, p50 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Detects endogenous levels of NFkB p105/p50 only when phosphorylated at serine 337. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| 1.0 mg/mL | |
| Western Blot 1:500, Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunofluorescence 1:100-1:200 | |
| P19838 | |
| NFKB1 | |
| The antiserum was produced against synthesized phosphopeptide derived from human NF-kB p105/p50 around the phosphorylation site of serine 337 (R-K-SP-D-L). | |
| 0.05 mL | |
| Apoptosis, Cancer, Cell Biology, Neurodegeneration, Neuroscience, Phospho Specific, Signal Transduction, Transcription Factors and Regulators | |
| 4790 | |
| Human, Mouse, Rat | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu