missing translation for 'onlineSavingsMsg'
Learn More

NFYC Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 10129-2-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16836313

  • 325.38€ / 150µL
Expédition estimée: 03-10-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The CCAAT-binding factor NF-Y is a heteromeric transcription factor that specifically binds to CCAAT sequences in many eukaryotic genes. NF-Y is made up of three subunits, NF-YA, NF-YB and NF-YC. All three components are necessary for DNA binding. In each NF-Y subunit, the segment needed for formation of the NF-Y-DNA complex is conserved from yeast to human. These conserved segments are homologous to the histone-fold motif of eukaryotic histones. The DNA binding domains of the NF-YB and NF-YC subunits have been suggested to interact through a protein-protein histone-fold "handshake" motif in a manner analogous to the histone proteins, H2B and H2A, respectively.
Spécification

Spécification

NFYC
Polyclonal
Unconjugated
NFYC
CBF C, CBFC, H1TF2A, HAP5, HSM, NF YC, NFYC, Transactivator HSM 1/2
Rabbit
Antigen Affinity Chromatography
RUO
18046, 25337, 4802
-20°C
Liquid
Western Blot
0.47 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P70353, Q13952, Q62725
Nfyc
NFYC Fusion Protein Ag0140
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions