missing translation for 'onlineSavingsMsg'
Learn More

Ninein Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Marque:  Novus Biologicals NBP2-97595-100ul

Nacres classification code: NA.46

 View more versions of this product

Product Code. 18687522

  • 326.00€ / 100µL

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

This item is not returnable. View return policy

Description

Description

Ninein Polyclonal antibody specifically detects Ninein in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Ninein
Polyclonal
Immunohistochemistry-Paraffin 1:50-1:200
Glycogen synthase kinase 3 beta-interacting protein, GSK3B-interacting protein, hNinein, KIAA1565, ninein, ninein (GSK3B interacting protein), ninein centrosomal protein
Produced in rabbits immunized with E. coli-derived Human Ninein fragment.
100 μg
Cell Biology, Cell Cycle and Replication
51199
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Immunohistochemistry (Paraffin)
Unconjugated
PBS (pH 7.0)
Rabbit
Antigen and protein A Affinity-purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.