missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOL12 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 15456-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
The protein encoded by this gene is a GTPase-activating protein (GAP) that associates with the Golgi apparatus and regulates the early secretory pathway of proteins. The encoded protein promotes hydrolysis of ADP-ribosylation factor 1 (ARF1)-bound GTP, which is required for the dissociation of coat proteins from Golgi-derived membranes and vesicles. Dissociation of the coat proteins is a prerequisite for the fusion of these vesicles with target compartments. The activity of this protein is sensitive to phospholipids. Multiple transcript variants encoding different isoforms have been found for this gene. This gene was originally known as ARFGAP1, but that is now the name of a related but different gene.Spécification
NOL12 | |
Polyclonal | |
Unconjugated | |
NOL12 | |
NOL12, Nop25, nucleolar protein 12 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
362955, 79159, 97961 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.24 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5D1Z3, Q8BG17, Q9UGY1 | |
Nol12 | |
NOL12 Fusion Protein Ag7723 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |