missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NR2E3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 14246-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
NR2E3, a NR2 Hepatocyte NF4-Like Receptor, has been suggested to affect cone cell proliferation in eye development. It has been shown to have a role in determining photoreceptor phenotype during human retinogenesis. In RD7 mice, a deletion in NR2E3 results in hereditary retinal dysplasia and degeneration. A homozygous missense mutation in NR2E3 has been linked to retinitis pigmentosa in Crypto-Jews of Belmonte. Two alternatively spliced isoforms have been documented. NR2E3 is expressed in the retina. ESTs have been isolated from cancerous human lung, colon, and ovary, and from normal human eye.Spécification
NR2E3 | |
Polyclonal | |
Unconjugated | |
NR2E3 | |
ESCS, NR2E3, PNR, rd7, RNR, RP37 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10002, 100365683, 23958 | |
-20°C | |
Liquid |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9QXZ7, Q9Y5X4 | |
Nr2e3 | |
NR2E3 Fusion Protein Ag5503 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |