Learn More
NSD3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marque: Invitrogen PA519427
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.01000kg
Description
PA5-19427 targets NSD3 in ICC/IF, IHC (P), and WB applications and shows reactivity with human samples. For IHC staining, heat mediated antigen retrieval is recommended. The PA5-19427 immunogen is synthetic peptide derived from residues 500 - 600 of Human NSD3. PA5-19427 detects NSD3 which has a predicted molecular weight of approximately 72.6 kDa and 161 kDa.
This gene is related to the Wolf-Hirschhorn syndrome candidate-1 gene and encodes a protein with PWWP domains. The function of the protein has not been determined. Two alternatively spliced variants have been described.Spécification
NSD3 | |
Polyclonal | |
Unconjugated | |
Q9BZ95 | |
NSD3 | |
Synthetic peptide derived from residues 500 - 600 of Human NSD3. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
NSD3 | |
DC28, DKFZp667H044, FLJ20353, MGC126766, MGC142029, NSD3, pp14328, WHSC1-like 1 isoform 9 with methyltransferase activity to lysine; WHSC1-like protein 1; histone-lysine N-methyltransferase NSD3; nuclear SET domain-containing protein 3; protein whistle | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54904 | |
-20°C or -80°C if preferred | |
Liquid |