Learn More
NSUN7 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 17546-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
 Afficher plus de versions de ce produit
Code produit. 16837454
  • 121.38€ / 20µL
Expédition estimée: 06-10-2022
pour voir le stock disponible 
Description
Description
The NSUN7 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human NSUN7. This antibody recognizes human, mouse, rat antigen. The NSUN7 antibody has been validated for the following applications: WB, ELISA analysis.
Spécification
Spécification
NSUN7
Polyclonal
Unconjugated
20 μL
79730
IgG
ELISA, Western Blot
233 μg/mL
Rabbit
Primary
Mouse, Rat, Human