missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUDT3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 20542-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
NUDT3 belongs to the MutT, or Nudix, protein family. Nudix proteins act as homeostatic checkpoints at important stages in nucleoside phosphate metabolic pathways, guarding against elevated levels of potentially dangerous intermediates, like 8-oxo-dGTP, which promotes AT-to-CG transversions (Safrany et al., 1998 ).Spécification
NUDT3 | |
Polyclonal | |
Unconjugated | |
NUDT3 | |
DIPP, DIPP 1, DIPP1, Nudix motif 3, NUDT3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11165, 56409 | |
-20°C | |
Liquid |
Western Blot | |
0.14 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O95989, Q9JI46 | |
Nudt3 | |
NUDT3 Fusion Protein Ag14587 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |