Learn More
NuMA Antibody (12A11), Novus Biologicals™
Rabbit Monoclonal Antibody
Marque: Novus Biologicals NBP3-26451-50ul
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
NuMA Monoclonal antibody specifically detects NuMA in Human samples. It is validated for Flow Cytometry, ELISA
Spécification
| NuMA | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA | |
| 12A11 | |
| Flow Cytometry 1:50-1:200, ELISA | |
| centrophilin stabilizes mitotic spindle in mitotic cells, nuclear mitotic apparatus protein 1, NUMA, NuMA protein, SP-H antigen, structural nuclear protein | |
| A synthesized peptide derived from Human NuMA [UniProt Q14980] | |
| 50 μL | |
| Breast Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Mitotic Regulators | |
| 4926 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |