missing translation for 'onlineSavingsMsg'
Learn More

NUP53 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 19819-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16829304

  • 325.38€ / 150µL
Expédition estimée: 06-10-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The elongation of primed DNA templates by DNA polymerase delta and DNA polymerase epsilon requires the accessory proteins proliferating cell nuclear antigen (PCNA) and replication factor C (RFC). RFC, also named activator 1, is a protein complex consisting of five distinct subunits of 140, 40, 38, 37, and 36 kDa. This gene encodes the 38 kDa subunit. This subunit is essential for the interaction between the 140 kDa subunit and the core complex that consists of the 36, 37, and 40 kDa subunits. Alternatively spliced transcript variants encoding distinct isoforms have been described.
Spécification

Spécification

NUP53
Polyclonal
Unconjugated
NUP35
35 kDa nucleoporin, Mitotic phosphoprotein 44, MP 44, MP44, nucleoporin 35kDa, Nucleoporin Nup35, Nucleoporin NUP53, NUP35, NUP53
Rabbit
Antigen Affinity Chromatography
RUO
129401, 295692, 69482
-20°C
Liquid
Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q68FY1, Q8NFH5, Q8R4R6
Nup35
NUP53 Fusion Protein Ag13869
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions