missing translation for 'onlineSavingsMsg'
Learn More

OCRL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 17695-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16837634

  • 140.00€ / 20µL
Expédition estimée: 12-04-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This antibody is specific to OCRL.

The inositol polyphosphate 5-phosphatases selectively remove the phosphate from the 5-position of various phosphatidylinositols, which generate second messengers in response to extracellular signals. OCRL1 is a type II 5-phosphatase that is mutated in the oculocerebrorenal syndrome of Lowe (OCRL). OCRL is a rare X-linked disorder that is characterized in part by congenital cataracts, mental retardation, muscular hypotonia and renal tubular dysfunction.OCRL1 has a high affinity for phosphatidylinositol 4,5-bisphosphate as well as inositol 1,4,5-trisphosphate and inositol 1,3,4,5-tetrakisphosphate as substrates. OCRL1 is localized to the Golgi complex and is thought to be part of the trans-Golgi network (TGN), which suggests that OCRL1 plays a role in protein sorting and trafficking within the cell.
Spécification

Spécification

OCRL
Polyclonal
Unconjugated
OCRL
INPP5F, LOCR, OCRL, OCRL1
Rabbit
Antigen Affinity Chromatography
RUO
317576, 320634, 4952
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
D3ZGS3, Q01968, Q6NVF0
Ocrl
OCRL Fusion Protein Ag11900
20 μL
Primary
Human, Rat, Mouse
Antibody
IgG
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions