missing translation for 'onlineSavingsMsg'
Learn More

ORAI3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 25766-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16894325

  • 349.00€ / 150µL
Expédition estimée: 09-12-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This antibody is predicted to have no cross-reactivity to ORAI1 or ORAI2. Larger molecular weight bands are sometimes seen in SDS-PAGE; these may represent post-translationally modified ORAI 3.

Antigen stimulation of immune cells triggers Ca++ entry t through Ca++ release-activated Ca++ (CRAC) channels. ORAI3 is one of two mammalian homologs to ORAI1, a recently identified four-transmembrane spanning protein that is an essential component of CRAC. All three homologs have been shown to function as Ca++ plasma membrane channels gated through interactions with STIM1, the store-activated endoplasmic reticulum Ca++ sensor. However, ORAI3 channels failed to produce detectable Ca++ selective currents in cells co-transfected with ORAI3 and STIM1, indicating that ORAI3 channels undergo a lesser degree of depotentiation than ORAI1 or ORAI2. Na+ currents through ORAI1, 2 and 3 channels were equally inhibited by extracellular Ca++, indicating that each have similar affinities for Ca++ within the selectivity filter. This antibody is predicted to have no cross-reactivity to ORAI1 or ORAI2. Larger molecular weight bands are sometimes seen in SDS-PAGE; these may represent post-translationally modified ORAI 3.
Spécification

Spécification

ORAI3
Polyclonal
Unconjugated
ORAI3
ORAI3, Protein orai 3, TMEM142C, Transmembrane protein 142C
Rabbit
Antigen Affinity Chromatography
RUO
269999, 93129
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q6P8G8, Q9BRQ5
ORAI3
ORAI3 Fusion Protein Ag22513
150 μL
Primary
Mouse, Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions