missing translation for 'onlineSavingsMsg'
Learn More

P-cadherin Mouse anti-Human, Clone: 1G10F7, Proteintech

Mouse Monoclonal Antibody

Marque:  Proteintech 66295-1-IG-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16888035

  • 399.00€ / 150µL
Expédition estimée: 29-04-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

P-cadherin, also known as Cadherin-3, is a Type 1 cadherin protein that belongs to the cadherin superfamily. Type 1 cadherins are single-pass transmembrane proteins that have 5 extracellular cadherin repeats and an intracellular domain that binds p120-catenin and beta-catenin. Cadherins are calcium-dependent cell-cell adhesion glycoproteins responsible for a range of processes including development, wound healing, cell-cell signaling, cell growth and differentiation. P-cadherin is expressed in human fetal structures, and in adult tissues such as the basal layer of the epidermis, breast, prostate, ovary, cervix, hair follicle, and corneal endothelium. P-cadherin expression has also been reported on embryonic stem cells, and stem cells of the normal mammary gland, and hair follicle. Overexpression of P-cadherin has been associated with poor prognosis in breast, prostate, ovary, colon, and stomach carcinomas. Mutations in CDH3 are associated with congential hypotrichosis with juvenile macular dystrophy.
Spécification

Spécification

P-cadherin
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P22223
CDH3
P-cadherin Fusion Protein Ag4758
150 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
1G10F7
Unconjugated
CDH3
Cadherin 3, CDH3, CDHP, HJMD, P cadherin, PCAD, P-cadherin, Placental cadherin
Mouse
Protein G
RUO
1001
-20°C
Liquid
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions