Learn More
P27 KIP1 Mouse anti-Human, Unconjugated, Clone: 3F12C10, Proteintech
Mouse Monoclonal Antibody
Marque:  Proteintech 67355-1-IG-20UL
Informations supplémentaires : Poids : 0.00010kg
 Afficher plus de versions de ce produit
Code produit. 16829945
  • 140.00€ / 20µL
Expédition estimée: 15-04-2024
pour voir le stock disponible 
Description
Description
The P27; KIP1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human P27; KIP1. This antibody recognizes human, mouse, rat antigen. The P27; KIP1 antibody has been validated for the following applications: WB, ELISA analysis.
Spécification
Spécification
P27 KIP1
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot
3F12C10
Unconjugated
20 μL
1027
IgG1