missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAH Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 16347-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
PAH encodes the enzyme phenylalanine hydroxylase that is the rate-limiting step in phenylalanine catabolism. Deficiency of this enzyme activity results in the autosomal recessive disorder phenylketonuria.Spécification
PAH | |
Polyclonal | |
Unconjugated | |
PAH | |
PAH, PH, Phe 4 monooxygenase, Phenylalanine 4 hydroxylase, phenylalanine hydroxylase, PKU, PKU1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18478, 24616, 5053 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.24 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P00439, P04176, P16331 | |
Pah | |
PAH Fusion Protein Ag9541 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |