missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAICS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 12967-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes a bifunctional enzyme containing phosphoribosylaminoimidazole carboxylase activity in its N-terminal region and phosphoribosylaminoimidazole succinocarboxamide synthetase in its C-terminal region. It catalyzes steps 6 and 7 of purine biosynthesis. The gene is closely linked and divergently transcribed with a locus that encodes an enzyme in the same pathway, and transcription of the two genes is coordinately regulated. The human genome contains several pseudogenes of this gene. Multiple transcript variants encoding different isoforms have been found for this gene.Spécification
PAICS | |
Polyclonal | |
Unconjugated | |
PAICS | |
ADE2, ADE2H1, AIR carboxylase, AIRC, PAICS, PAIS | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10606, 140946, 67054 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Flow Cytometry, Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P22234, P51583, Q9DCL9 | |
Paics | |
PAICS Fusion Protein Ag3621 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |