Learn More
Pancreatic Lipase Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 11209-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
This gene is a member of the lipase gene family. It encodes a carboxyl esterase that hydrolyzes insoluble, emulsified triglycerides, and is essential for the efficient digestion of dietary fats. This gene is expressed specifically in the pancreas.Spécification
| Pancreatic Lipase | |
| Polyclonal | |
| Unconjugated | |
| PNLIP | |
| Lipase, Pancreatic Lipase, PL, PNLIP, PTL | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 25702, 5406, 69060 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P16233, P27657, Q6P8U6 | |
| PNLIP | |
| Pancreatic Lipase Fusion Protein Ag1681 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |