missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Panendothelial Cell Antigen Antibody (MECA-32), DyLight 350, Novus Biologicals™
Rat Monoclonal Antibody
581.00€
Spécification
| Antigène | Panendothelial Cell Antigen |
|---|---|
| Clone | MECA-32 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugué | DyLight 350 |
Description
Panendothelial Cell Antigen Monoclonal specifically detects Panendothelial Cell Antigen in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Spécification
| Panendothelial Cell Antigen | |
| Western Blot | |
| DyLight 350 | |
| RUO | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| 84094 | |
| Mouse lymph node stromal cells. | |
| Primary | |
| Store at 4°C in the dark. |
| MECA-32 | |
| Monoclonal | |
| Rat | |
| Mouse | |
| MECA32, MECA-32, Panendothelial Cell Antigen | |
| Plvap | |
| IgG2a κ | |
| Affinity Purified | |
| Pan endothelial cells except for CNS & muscle capillaries. |
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit