Learn More
PARP1 Rabbit, anti-Human, Unconjugated, Clone: 3N19, Proteintech
Rabbit Recombinant Monoclonal Antibody
Marque:  Proteintech 80174-1-RR-20UL
Informations supplémentaires : Poids : 0.00020kg
 Afficher plus de versions de ce produit
Code produit. 17261546
  • 149.00€ / 20µL
Expédition estimée: 30-04-2024
pour voir le stock disponible 
Description
Description
The PARP1 antibody from Proteintech is a rabbit recombinant antibody to a recombinant protein of human PARP1. This antibody recognizes human, mouse, rat antigen. The PARP1 antibody has been validated for the following applications: WB,ELISA analysis.
Spécification
Spécification
PARP1
Monoclonal
250 μg/mL
Rabbit
Primary
Human, Mouse, Rat
Western Blot, ELISA
3N19
Unconjugated
20 μL
142