Learn More
PAX8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 10336-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
This antibody can detect all 5 isoforms of PAX8 (31 kDa, 34 kDa, 41 kDa, 43 kDa, and 48 kDa). A 58-60 kDa form has also been reported. Proteintech's 10336-1-AP antibody has cross-reactivities with the other PAX family members.
PAX8 is a member of the paired box (PAX) family of transcription factors. Members of this gene family typically contain a paired box domain, an octapeptide, and a paired-type homeodomain. Pax8 is expressed in the developing and adult thyroid and kidney. In the thyroid, PAX8 regulates thyroglobulin, thyroidperoxidase,and NIS (sodium/iodide symporter) expression. Mutations in PAX8 have been associated with thyroid dysgenesis, thyroid follicular carcinomas and atypical follicular thyroid adenomas.Spécification
PAX8 | |
Polyclonal | |
Unconjugated | |
PAX8 | |
paired box 8, Paired box protein Pax 8, PAX8 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18510, 7849, 81819 | |
-20°C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P51974, Q00288, Q06710 | |
Pax8 | |
PAX8 Fusion Protein Ag0306 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |