Learn More
PCF11 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 23540-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
This antibody is specific to the 173 kDa human PCF11 protein.
Component of pre-mRNA cleavage complex II.Spécification
PCF11 | |
Polyclonal | |
Unconjugated | |
PCF11 | |
KIAA0824, PCF11 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
51585 | |
-20°C | |
Liquid |
Western Blot | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O94913 | |
PCF11 | |
PCF11 Fusion Protein Ag20263 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |