missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCYOX1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 18868-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
Prenylcysteine is released during the degradation of prenylated proteins. PCYOX1 catalyzes the degradation of prenylcysteine to yield free cysteines and a hydrophobic isoprenoid product (Tschantz et al., 1999 ).Spécification
PCYOX1 | |
Polyclonal | |
Unconjugated | |
PCYOX1 | |
KIAA0908, PCL1, PCYOX1, Prenylcysteine lyase, prenylcysteine oxidase 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
246302, 51449, 66881 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q99ML5, Q9CQF9, Q9UHG3 | |
PCYOX1 | |
PCYOX1 Fusion Protein Ag4316 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |