missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDCD4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 12587-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes a protein localized to the nucleus in proliferating cells. Expression of this gene is modulated by cytokines in natural killer and T cells. The gene product is thought to play a role in apoptosis but the specific role has not yet been determined. Two transcripts encoding different isoforms have been identified.Spécification
PDCD4 | |
Polyclonal | |
Unconjugated | |
PDCD4 | |
H731, Nuclear antigen H731 like, PDCD4, Protein 197/15a | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18569, 27250, 64031 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunohistochemistry (Paraffin), Flow Cytometry, Western Blot | |
0.6 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q53EL6, Q61823, Q9JID1 | |
PDCD4 | |
PDCD4 Fusion Protein Ag3288 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |