Learn More
PDCL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 16057-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Code produit. 16835534
  • 121.38€ / 20µL
Expédition estimée: 30-09-2022
Description
Phosducin-like protein is a putative modulator of heterotrimeric G proteins. The protein shares extensive amino acid sequence homology with phosducin, a phosphoprotein expressed in retina and pineal gland. Both phosducin-like protein and phosphoducin have been shown to regulate G-protein signaling by binding to the beta-gamma subunits of G proteins.
Spécification
Spécification
PDCL
Polyclonal
Unconjugated
PDCL
PDCL, PhLP, phosducin like, Phosducin like protein
Rabbit
Antigen Affinity Chromatography
RUO
5082, 64013, 67466
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.25 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q13371, Q63737, Q9DBX2
PDCL
PDCL Fusion Protein Ag9029
20 μL
Primary
Human, Mouse, Rat