missing translation for 'onlineSavingsMsg'
Learn More

Phospho-PLK1 (Ser326) Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 19553-1-AP-100UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16829094

  • 325.38€ / 100µL
Expédition estimée: 30-09-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The antibody is specific to Ser(326)-phospho.

PLK1 (Polo Like Kinase 1) is a Protein Coding gene. Diseases associated with PLK1 include Transient Tic Disorder and Monoclonal Paraproteinemia. Among its related pathways are Integrated Breast Cancer Pathway and Cell Cycle, Mitotic. Gene Ontology (GO) annotations related to this gene include transferase activity, transferring phosphorus-containing groups and protein tyrosine kinase activity. An important paralog of this gene is PLK3. This gene encodes a protein which contains a conserved nuclear localization signal.
Spécification

Spécification

Phospho-PLK1 (Ser326)
Polyclonal
Unconjugated
PLK1
PLK, PLK 1, PLK1, Polo like kinase 1, STPK13
Rabbit
Antigen Affinity Chromatography
RUO
18817, 25515, 5347
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P53350, Q07832, Q62673
Plk1
Peptide
100 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions